Product Details

SNP ID
rs147449402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101792866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTTAGGAAAATAAGGTCTGAT[A/C]GTATAAACTTTGGAGGTAGGAGTCA
Phenotype
MIM: 604039 MIM: 606456
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MGEA5 PubMed Links

Gene Details

Gene
MGEA5
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142434.1 2968 Silent Mutation ACG,ACT T663T NP_001135906.1
NM_012215.3 2968 Silent Mutation ACG,ACT T716T NP_036347.1
XM_017015585.1 2968 Silent Mutation ACG,ACT T601T XP_016871074.1
XM_017015586.1 2968 Silent Mutation ACG,ACT T578T XP_016871075.1
Gene
NPM3
Gene Name
nucleophosmin/nucleoplasmin 3
There are no transcripts associated with this gene.

View Full Product Details