Product Details

SNP ID
rs147488920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15213008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTCATCATCATCATCGTCTTC[C/T]TCTTCCTCGTGGGCAGATCTTCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM171A1 PubMed Links

Gene Details

Gene
FAM171A1
Gene Name
family with sequence similarity 171 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010924.1 2508 Silent Mutation GAA,GAG E860E NP_001010924.1
XM_011519378.2 2508 Silent Mutation GAA,GAG E847E XP_011517680.1
XM_017015904.1 2508 Silent Mutation GAA,GAG E831E XP_016871393.1
XM_017015905.1 2508 Silent Mutation GAA,GAG E502E XP_016871394.1

View Full Product Details