Product Details

SNP ID
rs147615429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112286077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCTTTCTTTGTCCAGTACAAG[A/C]CACCTATCTATCACTTCTACAGTCA
Phenotype
MIM: 602653
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TECTB PubMed Links

Gene Details

Gene
TECTB
Gene Name
tectorin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058222.2 347 Missense Mutation ACA,CCA T92P NP_478129.1
XM_017016604.1 347 Missense Mutation ACA,CCA T92P XP_016872093.1

View Full Product Details