Product Details
- SNP ID
-
rs147639368
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:100042522 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGACTCCGTGCCTTCTGCTGGGAG[A/C]TCTCCTCACAGGGCTTACTTGAGGG
- Phenotype
-
MIM: 603103
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPN1
PubMed Links
Gene Details
- Gene
- CPN1
- Gene Name
- carboxypeptidase N subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308.2 |
1542 |
Missense Mutation |
GCT,TCT |
A428S |
NP_001299.1 |
View Full Product Details