Product Details

SNP ID
rs147757005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91152511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGAGGTGACCCTCCCTGTCCT[A/G]TTTAGTGGGAGTTATGTGCTGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT9P1 PubMed Links

Gene Details

Gene
NUDT9P1
Gene Name
nudix hydrolase 9 pseudogene 1
There are no transcripts associated with this gene.

Gene
PCGF5
Gene Name
polycomb group ring finger 5
There are no transcripts associated with this gene.

View Full Product Details