Product Details

SNP ID
rs147861232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68960044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTGAAAAGAAAGTGGTTTCTT[C/T]TAAAACCAAAAAAGTGACAAAAAAT
Phenotype
MIM: 606357 MIM: 610373
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX21 PubMed Links

Gene Details

Gene
DDX21
Gene Name
DEAD-box helicase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256910.1 548 Missense Mutation TCT,TTT S41F NP_001243839.1
NM_004728.3 548 Missense Mutation TCT,TTT S109F NP_004719.2
XM_011540336.1 548 Missense Mutation TCT,TTT S109F XP_011538638.1
XM_017016910.1 548 Missense Mutation TCT,TTT S41F XP_016872399.1
Gene
DDX50
Gene Name
DEAD-box helicase 50
There are no transcripts associated with this gene.

View Full Product Details