Product Details

SNP ID
rs147914123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43644783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCAGAGTGGTCATATTTGTGG[C/T]GGGCTTCAGTCATCACATCTGATAA
Phenotype
MIM: 194539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF32 PubMed Links

Gene Details

Gene
ZNF32
Gene Name
zinc finger protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005368.2 235 Missense Mutation CAC,CGC H30R NP_001005368.1
NM_001324164.1 235 Missense Mutation CAC,CGC H34R NP_001311093.1
NM_001324165.1 235 Missense Mutation CAC,CGC H34R NP_001311094.1
NM_001324166.1 235 Missense Mutation CAC,CGC H5R NP_001311095.1
NM_001324167.1 235 Missense Mutation CAC,CGC H5R NP_001311096.1
NM_001324168.1 235 Missense Mutation CAC,CGC H5R NP_001311097.1
NM_006973.2 235 Missense Mutation CAC,CGC H30R NP_008904.1
XM_017016616.1 235 Missense Mutation CAC,CGC H30R XP_016872105.1
Gene
ZNF32-AS1
Gene Name
ZNF32 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF32-AS2
Gene Name
ZNF32 antisense RNA 2
There are no transcripts associated with this gene.

Gene
ZNF32-AS3
Gene Name
ZNF32 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details