Product Details

SNP ID
rs148029829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5393818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACACCAATCTACAAACTGAACA[C/G]AGTGCCTCGACTTCGTGGCTGCGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TUBAL3 PubMed Links

Gene Details

Gene
TUBAL3
Gene Name
tubulin alpha like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171864.1 1078 Missense Mutation TCT,TGT S307C NP_001165335.1
NM_024803.2 1078 Missense Mutation TCT,TGT S347C NP_079079.1

View Full Product Details