Product Details

SNP ID
rs148315007
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23440579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCGCTGCTGCCCAAGACGGGGC[A/T]TGGGCCGGGTACCCGGAGTTGCTGG
Phenotype
MIM: 612022
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
OTUD1 PubMed Links

Gene Details

Gene
OTUD1
Gene Name
OTU deubiquitinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145373.2 1311 Silent Mutation GCA,GCT A374A NP_001138845.1

View Full Product Details