Product Details

SNP ID
rs148626202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16825009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTTAATCGGAATGCGGATGGA[C/T]GCCGTGCATAATCAGCATGAAATTT
Phenotype
MIM: 602997 MIM: 179555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUBN PubMed Links

Gene Details

Gene
CUBN
Gene Name
cubilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081.3 7253 Missense Mutation CAT,CGT H3613R NP_001072.2
XM_011519708.2 7253 Intron XP_011518010.1
XM_011519709.2 7253 Missense Mutation CAT,CGT H2275R XP_011518011.1
XM_011519710.2 7253 Missense Mutation CAT,CGT H2267R XP_011518012.1
XM_011519711.2 7253 Missense Mutation CAT,CGT H2227R XP_011518013.1
Gene
RSU1
Gene Name
Ras suppressor protein 1
There are no transcripts associated with this gene.

View Full Product Details