Product Details

SNP ID
rs148806171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45003890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCAGTGCAGTGAATGTGGCAA[A/G]TGTTTCAGTCAGAGCTCTCATCTGA
Phenotype
MIM: 610559 MIM: 194529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf25 PubMed Links

Gene Details

Gene
C10orf25
Gene Name
chromosome 10 open reading frame 25
There are no transcripts associated with this gene.

Gene
RASSF4
Gene Name
Ras association domain family member 4
There are no transcripts associated with this gene.

Gene
ZNF22
Gene Name
zinc finger protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006963.4 765 Silent Mutation AAA,AAG K174K NP_008894.2

View Full Product Details