Product Details

SNP ID
rs149008935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100978305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTGTTCCCTAGGATGCTGAGG[A/C]CTTCACCTTGCCAACCAGCTTCGAG
Phenotype
MIM: 606075 MIM: 611848
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf2 PubMed Links

Gene Details

Gene
C10orf2
Gene Name
chromosome 10 open reading frame 2
There are no transcripts associated with this gene.

Gene
MIR608
Gene Name
microRNA 608
There are no transcripts associated with this gene.

Gene
MRPL43
Gene Name
mitochondrial ribosomal protein L43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308396.1 1668 Intron NP_001295325.1
NM_032112.2 1668 Intron NP_115488.2
NM_176792.2 1668 UTR 3 NP_789762.1
NM_176793.1 1668 Intron NP_789763.1
NM_176794.1 1668 Intron NP_789764.1
XM_005270231.2 1668 Intron XP_005270288.1
XM_006718035.3 1668 Intron XP_006718098.1
XM_017016790.1 1668 Intron XP_016872279.1
Gene
SEMA4G
Gene Name
semaphorin 4G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001203244.1 1668 Missense Mutation GAC,GCC D149A NP_001190173.1
NM_017893.3 1668 Missense Mutation GAC,GCC D149A NP_060363.2
XM_005270008.2 1668 Missense Mutation GAC,GCC D149A XP_005270065.1

View Full Product Details