Product Details
- SNP ID
-
hCV160804615
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5393720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTTGCTCAGCATGCAGATGGACC[A/G]GTGGACTTTGGCCAGGTCCCCACCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TUBAL3
PubMed Links
Gene Details
- Gene
- TUBAL3
- Gene Name
- tubulin alpha like 3
There are no transcripts associated with this gene.
View Full Product Details