Product Details

SNP ID
rs149161234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117201190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACAGAAAACAGGATCTCCAGGG[C/G]CATCTGCAGAAGGTGAAGCCTCAGT
Phenotype
MIM: 613655
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNK18 PubMed Links

Gene Details

Gene
KCNK18
Gene Name
potassium two pore domain channel subfamily K member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181840.1 255 Silent Mutation GGC,GGG G85G NP_862823.1

View Full Product Details