Product Details
- SNP ID
-
rs149276130
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5525151 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGCCTTCTCCCTGTTTGACAA[A/G]GATGGGGACGGCTGCATCACCACCC
- Phenotype
-
MIM: 114184
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CALML3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2231420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CALML3
- Gene Name
- calmodulin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005185.3 |
263 |
Silent Mutation |
AAA,AAG |
K22K |
NP_005176.1 |
- Gene
- CALML3-AS1
- Gene Name
- CALML3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details