Product Details

SNP ID
rs149475857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103476351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCGAGCAGGGCTATCCCTGAC[C/T]AGCTCATCCTCCTTGCAGGGAACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALHM3 PubMed Links

Gene Details

Gene
CALHM3
Gene Name
calcium homeostasis modulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129742.1 694 Silent Mutation NP_001123214.1
Gene
NEURL1-AS1
Gene Name
NEURL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details