Product Details

SNP ID
rs149480041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103967844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGGACCTGAACCCCGAAGACTT[G/T]TGGGAGATTATAGGAGAACTGGGCG
Phenotype
MIM: 616563
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLK PubMed Links

Gene Details

Gene
SLK
Gene Name
STE20 like kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304743.1 672 Silent Mutation TTG,TTT L33F NP_001291672.1
NM_014720.3 672 Silent Mutation TTG,TTT L33F NP_055535.2
XM_011540401.2 672 Missense Mutation TTG,TTT L33F XP_011538703.1

View Full Product Details