Product Details

SNP ID
rs149719070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5693090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTAACGTTTTACCTTCTGGTGAG[G/T]GTGTGGGGAGAGAAGCTTTGGCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM208B PubMed Links

Gene Details

Gene
FAM208B
Gene Name
family with sequence similarity 208 member B
There are no transcripts associated with this gene.

View Full Product Details