Product Details

SNP ID
rs149741269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1024681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCAGCATTTCCTCCAAGCGCT[A/G]CAACTGACGCCTGTCAACCCAGTCA
Phenotype
MIM: 615389 MIM: 615391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP4 PubMed Links

Gene Details

Gene
GTPBP4
Gene Name
GTP binding protein 4
There are no transcripts associated with this gene.

Gene
IDI2
Gene Name
isopentenyl-diphosphate delta isomerase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033261.2 108 Nonsense Mutation CAG,TAG Q15* NP_150286.1
Gene
IDI2-AS1
Gene Name
IDI2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details