Product Details

SNP ID
rs149817075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5895131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCAGGGCCCCTGGGCCATCTCC[A/G]CAGGAGATGCCAGAGGACGAGTGCC
Phenotype
MIM: 607222
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD16 PubMed Links

Gene Details

Gene
ANKRD16
Gene Name
ankyrin repeat domain 16
There are no transcripts associated with this gene.

Gene
FBXO18
Gene Name
F-box protein, helicase, 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258452.1 611 Intron NP_001245381.1
NM_001258453.1 611 Intron NP_001245382.1
NM_032807.4 611 Missense Mutation CAC,CGC H33R NP_116196.3
NM_178150.2 611 Intron NP_835363.1
XM_011519748.2 611 Missense Mutation CAC,CGC H33R XP_011518050.1
XM_017016842.1 611 Missense Mutation CAC,CGC H33R XP_016872331.1
XM_017016843.1 611 Missense Mutation CAC,CGC H33R XP_016872332.1
XM_017016844.1 611 Intron XP_016872333.1
XM_017016845.1 611 Missense Mutation CAC,CGC H33R XP_016872334.1
XM_017016846.1 611 Intron XP_016872335.1

View Full Product Details