Product Details

SNP ID
rs149964494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122833678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATAGCTTTTTTCTTACCTGAAT[G/T]GCCACTTGCACTTCGATCCCTTTTC
Phenotype
MIM: 616644
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CUZD1 PubMed Links

Gene Details

Gene
CUZD1
Gene Name
CUB and zona pellucida like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022034.5 1977 Missense Mutation AAT,CAT N549H NP_071317.2
Gene
FAM24B-CUZD1
Gene Name
FAM24B-CUZD1 readthrough
There are no transcripts associated with this gene.

Gene
LOC729800
Gene Name
deleted in malignant brain tumors 1 protein
There are no transcripts associated with this gene.

View Full Product Details