Product Details

SNP ID
rs150311863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:47366973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGCTTCACCAAGGGCAGCCTA[T/C]ACTTTTCCACTCCTTAATATTGGGC
Phenotype
MIM: 180290
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RBP3 PubMed Links

Gene Details

Gene
RBP3
Gene Name
retinol binding protein 3
There are no transcripts associated with this gene.

Gene
ZNF488
Gene Name
zinc finger protein 488
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153034.2 2068 UTR 3 NP_694579.1
XM_006717617.3 2068 UTR 3 XP_006717680.1
XM_011539244.2 2068 UTR 3 XP_011537546.1
XM_017015642.1 2068 UTR 3 XP_016871131.1
XM_017015643.1 2068 UTR 3 XP_016871132.1

View Full Product Details