Product Details

SNP ID
rs150521753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101611638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGACACTTACGTGCAGGCAG[G/T]CCCTTTGACGCCGGTCCCACAGCCG
Phenotype
MIM: 616467 MIM: 608071
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
FBXW4
Gene Name
F-box and WD repeat domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323541.1 Intron NP_001310470.1
NM_022039.3 Intron NP_071322.1
XM_005270053.2 Intron XP_005270110.2
XM_017016570.1 Intron XP_016872059.1

View Full Product Details