Product Details

SNP ID
rs150532542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125824871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTAGATGGCTGGATCAGAAT[A/C]AGGAAGAGAGAGTGATAGGAAATAA
Phenotype
MIM: 611883 MIM: 606938
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCCIP PubMed Links

Gene Details

Gene
BCCIP
Gene Name
BRCA2 and CDKN1A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016567.3 Intron NP_057651.1
NM_078468.2 Intron NP_510868.1
NM_078469.2 Intron NP_510869.1
Gene
MIR4484
Gene Name
microRNA 4484
There are no transcripts associated with this gene.

Gene
UROS
Gene Name
uroporphyrinogen III synthase
There are no transcripts associated with this gene.

View Full Product Details