Product Details
- SNP ID
-
rs150655034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124623260 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCCCTCAGTTCTTCTCTATCTG[C/T]TCAATGTCCAACTCGCCGTCCAGGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM53B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139871474] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM53B
- Gene Name
- family with sequence similarity 53 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014661.3 |
1774 |
Silent Mutation |
GAA,GAG |
E417E |
NP_055476.3 |
- Gene
- LHPP
- Gene Name
- phospholysine phosphohistidine inorganic pyrophosphate phosphatase
There are no transcripts associated with this gene.
View Full Product Details