Product Details

SNP ID
rs150863046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84245133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCACTGGCTTCGGGGAGCTC[A/G]AGGTGCTGGCTGTGGGGATGGTGCT
Phenotype
MIM: 616103 MIM: 600342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRIT1 PubMed Links

Gene Details

Gene
LRIT1
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
There are no transcripts associated with this gene.

Gene
RGR
Gene Name
retinal G protein coupled receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012720.1 81 Missense Mutation AAG,GAG K15E NP_001012738.1
NM_001012722.1 81 Missense Mutation AAG,GAG K15E NP_001012740.1
NM_002921.3 81 Missense Mutation AAG,GAG K15E NP_002912.2

View Full Product Details