Product Details

SNP ID
rs150903081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5870334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCACCCGCACAGTGAGGTCAG[C/T]GTTGGCTCTTGGCCTCTCTTCCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD16 PubMed Links

Gene Details

Gene
ANKRD16
Gene Name
ankyrin repeat domain 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009941.2 Intron NP_001009941.1
NM_001009943.2 Intron NP_001009943.1
NM_019046.2 Intron NP_061919.1

View Full Product Details