Product Details

SNP ID
rs151036418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102726739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTAAACATCACGGACCCCCGC[A/G]CTGTCTTTGTATCTGAGCGGGAGCT
Phenotype
MIM: 615570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFXN2 PubMed Links

Gene Details

Gene
SFXN2
Gene Name
sideroflexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178858.4 317 Missense Mutation ACT,GCT T35A NP_849189.1
XM_011539261.2 317 Missense Mutation ACT,GCT T44A XP_011537563.1
XM_011539262.2 317 Missense Mutation ACT,GCT T35A XP_011537564.1
XM_011539263.2 317 UTR 5 XP_011537565.1
XM_011539264.2 317 UTR 5 XP_011537566.1
XM_017015663.1 317 Missense Mutation ACT,GCT T53A XP_016871152.1

View Full Product Details