Product Details

SNP ID
rs151085716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80078761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTCTGTTCTGGTTCACAGTCA[A/T]CACCCTCAGCTTTGGCTACTACACA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM254 PubMed Links

Gene Details

Gene
TMEM254
Gene Name
transmembrane protein 254
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270367.1 116 Intron NP_001257296.1
NM_001270368.1 116 Intron NP_001257297.1
NM_001270369.1 116 Intron NP_001257298.1
NM_001270370.1 116 Missense Mutation AAC,ATC N21I NP_001257299.1
NM_001270371.1 116 Missense Mutation AAC,ATC N21I NP_001257300.1
NM_001270372.1 116 Missense Mutation AAC,ATC N21I NP_001257301.1
NM_001270373.1 116 Missense Mutation AAC,ATC N21I NP_001257302.1
NM_001270374.1 116 Intron NP_001257303.1
NM_025125.3 116 Missense Mutation AAC,ATC N21I NP_079401.2
XM_006717978.2 116 Intron XP_006718041.1
XM_011540192.2 116 Intron XP_011538494.1
XM_011540193.2 116 Intron XP_011538495.1
XM_017016679.1 116 Intron XP_016872168.1
XM_017016680.1 116 Intron XP_016872169.1
Gene
TMEM254-AS1
Gene Name
TMEM254 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details