Product Details
- SNP ID
-
rs151237508
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:68341309 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTCTAAAGATAAAAATAACATG[C/T]GTAAGTGGTGTCTTTGGCACACAAT
- Phenotype
-
MIM: 602324
MIM: 612189
MIM: 610328
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPH3
PubMed Links
Gene Details
- Gene
- HNRNPH3
- Gene Name
- heterogeneous nuclear ribonucleoprotein H3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322434.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q259* |
NP_001309363.1 |
NM_001322436.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q259* |
NP_001309365.1 |
NM_001322437.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q259* |
NP_001309366.1 |
NM_001322438.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q244* |
NP_001309367.1 |
NM_001322439.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q244* |
NP_001309368.1 |
NM_001322440.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q244* |
NP_001309369.1 |
NM_001322441.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q244* |
NP_001309370.1 |
NM_001322442.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q253* |
NP_001309371.1 |
NM_001322443.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q253* |
NP_001309372.1 |
NM_001322444.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q151* |
NP_001309373.1 |
NM_001322445.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
NP_001309374.1 |
NM_001322446.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
NP_001309375.1 |
NM_001322447.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
NP_001309376.1 |
NM_001322448.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
NP_001309377.1 |
NM_001322449.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
NP_001309378.1 |
NM_001322450.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q122* |
NP_001309379.1 |
NM_001322451.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q90* |
NP_001309380.1 |
NM_001322452.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q90* |
NP_001309381.1 |
NM_001322453.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q90* |
NP_001309382.1 |
NM_012207.2 |
986 |
Nonsense Mutation |
CAA,TAA |
Q259* |
NP_036339.1 |
NM_021644.3 |
986 |
Nonsense Mutation |
CAA,TAA |
Q244* |
NP_067676.2 |
XM_011539742.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q151* |
XP_011538044.1 |
XM_017016168.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q259* |
XP_016871657.1 |
XM_017016169.1 |
986 |
Nonsense Mutation |
CAA,TAA |
Q128* |
XP_016871658.1 |
- Gene
- PBLD
- Gene Name
- phenazine biosynthesis like protein domain containing
There are no transcripts associated with this gene.
- Gene
- RUFY2
- Gene Name
- RUN and FYVE domain containing 2
View Full Product Details