Product Details

SNP ID
rs111363197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63506559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACTCTAATGTGGAACCTCCTC[A/G]GTCTCTGGGCTCTTCCCTTCCAACT
Phenotype
MIM: 606096
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGALS12 PubMed Links

Gene Details

Gene
LGALS12
Gene Name
galectin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142535.1 Intron NP_001136007.1
NM_001142536.1 Intron NP_001136008.1
NM_001142537.1 Intron NP_001136009.1
NM_001142538.1 Intron NP_001136010.1
NM_033101.3 Intron NP_149092.2
XM_017018458.1 Intron XP_016873947.1

View Full Product Details