Product Details

SNP ID
rs111687572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67292034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGTGGACCATGACCGGCAGGT[A/G]GTGCATGTGGAGACACTGGGGCTCA
Phenotype
MIM: 615126 MIM: 109635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD13D PubMed Links

Gene Details

Gene
ANKRD13D
Gene Name
ankyrin repeat domain 13D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207354.2 18 Silent Mutation GTA,GTG V195V NP_997237.2
XM_011544985.1 18 Silent Mutation GTA,GTG V195V XP_011543287.1
XM_011544986.1 18 Silent Mutation GTA,GTG V108V XP_011543288.1
XM_011544987.2 18 Silent Mutation GTA,GTG V108V XP_011543289.1
XM_011544988.2 18 UTR 5 XP_011543290.1
Gene
GRK2
Gene Name
G protein-coupled receptor kinase 2
There are no transcripts associated with this gene.

View Full Product Details