Product Details

SNP ID
rs112713208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73309520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCGCTGGGGAGGGAGGCCCGG[C/G]CTCAGGCCTGGAAGCTCCCTATTGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF17 PubMed Links

Gene Details

Gene
ARHGEF17
Gene Name
Rho guanine nucleotide exchange factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014786.3 903 Silent Mutation GGC,GGG G294G NP_055601.2
XM_017018623.1 903 Intron XP_016874112.1
XM_017018624.1 903 Intron XP_016874113.1

View Full Product Details