Product Details

SNP ID
rs112893496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65041589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCGCCCGCGCCGAGGTGGCCAG[C/T]TTCGTGGCAGACCGCTTGCGAGCTG
Phenotype
MIM: 602640 MIM: 605964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL2-SNX15 PubMed Links

Gene Details

Gene
ARL2-SNX15
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
NAALADL1
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
There are no transcripts associated with this gene.

Gene
SAC3D1
Gene Name
SAC3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013299.3 686 Silent Mutation AGC,AGT S99S NP_037431.3
Gene
SNX15
Gene Name
sorting nexin 15
There are no transcripts associated with this gene.

View Full Product Details