Product Details
- SNP ID
-
rs112998492
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59457204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCATCTATCACCGTCCCCAAGTTC[C/T]TGGTGGATCTTTTATCAGACAGGAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4D6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1453543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4D6
- Gene Name
- olfactory receptor family 4 subfamily D member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004708.1 |
244 |
Silent Mutation |
CTG,TTG |
L82L |
NP_001004708.1 |
View Full Product Details