Product Details
- SNP ID
-
rs113420599
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74843877 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTAGTGGAGCCTCTGTAATGTTC[C/T]GGGGATCCCGCAAGCGAATGAAGAT
- Phenotype
-
MIM: 609788
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF169
PubMed Links
Gene Details
- Gene
- RNF169
- Gene Name
- ring finger protein 169
There are no transcripts associated with this gene.
- Gene
- XRRA1
- Gene Name
- X-ray radiation resistance associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001270380.1 |
2172 |
Intron |
|
|
NP_001257309.1 |
NM_001270381.1 |
2172 |
Missense Mutation |
CCG,CTG |
P426L |
NP_001257310.1 |
NM_182969.2 |
2172 |
Missense Mutation |
CCG,CTG |
P701L |
NP_892014.1 |
XM_005273765.2 |
2172 |
Missense Mutation |
CCG,CTG |
P709L |
XP_005273822.1 |
XM_005273770.2 |
2172 |
Missense Mutation |
CCG,CTG |
P468L |
XP_005273827.1 |
XM_011544755.1 |
2172 |
Missense Mutation |
CCG,CTG |
P694L |
XP_011543057.1 |
XM_011544756.1 |
2172 |
Missense Mutation |
CCG,CTG |
P667L |
XP_011543058.1 |
XM_011544757.1 |
2172 |
Missense Mutation |
CCG,CTG |
P652L |
XP_011543059.1 |
XM_011544758.1 |
2172 |
Missense Mutation |
CCG,CTG |
P652L |
XP_011543060.1 |
XM_011544759.1 |
2172 |
Missense Mutation |
CCG,CTG |
P644L |
XP_011543061.1 |
XM_011544760.2 |
2172 |
Missense Mutation |
CCG,CTG |
P709L |
XP_011543062.1 |
XM_011544761.1 |
2172 |
Missense Mutation |
CCG,CTG |
P628L |
XP_011543063.1 |
XM_011544762.1 |
2172 |
Missense Mutation |
CCG,CTG |
P622L |
XP_011543064.1 |
XM_011544763.1 |
2172 |
Intron |
|
|
XP_011543065.1 |
XM_011544764.1 |
2172 |
Missense Mutation |
CCG,CTG |
P597L |
XP_011543066.1 |
XM_011544765.1 |
2172 |
Missense Mutation |
CCG,CTG |
P468L |
XP_011543067.1 |
XM_011544766.2 |
2172 |
Missense Mutation |
CCG,CTG |
P546L |
XP_011543068.1 |
XM_011544767.1 |
2172 |
Missense Mutation |
CCG,CTG |
P546L |
XP_011543069.1 |
XM_011544768.1 |
2172 |
Missense Mutation |
CCG,CTG |
P546L |
XP_011543070.1 |
XM_017017221.1 |
2172 |
Missense Mutation |
CCG,CTG |
P639L |
XP_016872710.1 |
XM_017017222.1 |
2172 |
Intron |
|
|
XP_016872711.1 |
XM_017017223.1 |
2172 |
Intron |
|
|
XP_016872712.1 |
XM_017017224.1 |
2172 |
Intron |
|
|
XP_016872713.1 |
XM_017017225.1 |
2172 |
Intron |
|
|
XP_016872714.1 |
XM_017017226.1 |
2172 |
Missense Mutation |
CCG,CTG |
P468L |
XP_016872715.1 |
XM_017017227.1 |
2172 |
Missense Mutation |
CCG,CTG |
P468L |
XP_016872716.1 |
XM_017017228.1 |
2172 |
Intron |
|
|
XP_016872717.1 |
XM_017017229.1 |
2172 |
Missense Mutation |
CCG,CTG |
P411L |
XP_016872718.1 |
XM_017017230.1 |
2172 |
Missense Mutation |
CCG,CTG |
P389L |
XP_016872719.1 |
XM_017017231.1 |
2172 |
Missense Mutation |
CCG,CTG |
P381L |
XP_016872720.1 |
XM_017017232.1 |
2172 |
Intron |
|
|
XP_016872721.1 |
XM_017017233.1 |
2172 |
Missense Mutation |
CCG,CTG |
P324L |
XP_016872722.1 |
XM_017017234.1 |
2172 |
Missense Mutation |
CCG,CTG |
P324L |
XP_016872723.1 |
View Full Product Details