Product Details

SNP ID
rs113543168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66332380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAGCAGTTGTCTCAGACTGTT[C/G]CCGAATGTCCCTGGGGCTGGCTTTG
Phenotype
MIM: 606259 MIM: 605965
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRMS1 PubMed Links

Gene Details

Gene
BRMS1
Gene Name
breast cancer metastasis suppressor 1
There are no transcripts associated with this gene.

Gene
RIN1
Gene Name
Ras and Rab interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004292.2 2724 Missense Mutation CAA,GAA Q750E NP_004283.2
XM_011545399.2 2724 Missense Mutation CAA,GAA Q722E XP_011543701.1
XM_011545400.2 2724 Missense Mutation CAA,GAA Q688E XP_011543702.1
XM_011545401.2 2724 Missense Mutation CAA,GAA Q669E XP_011543703.1
XM_017018587.1 2724 Missense Mutation CAA,GAA Q689E XP_016874076.1

View Full Product Details