Product Details

SNP ID
rs113911443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56290519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGCAGGACAGAGCTAAAATTG[A/G]AGACGTGTCGCAGAAAAAGTGACGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8H1 PubMed Links

Gene Details

Gene
OR8H1
Gene Name
olfactory receptor family 8 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005199.1 Intron NP_001005199.1

View Full Product Details