Product Details

SNP ID
rs114087729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67662676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCCCAGTTTTCCCAGTGGATGGA[A/G]CTGCTGTGGGTGAGTGCAGGGTGCG
Phenotype
MIM: 601917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3B2 PubMed Links

Gene Details

Gene
ALDH3B2
Gene Name
aldehyde dehydrogenase 3 family member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000695.3 1980 UTR 3 NP_000686.2
NM_001031615.1 1980 UTR 3 NP_001026786.1

View Full Product Details