Product Details

SNP ID
rs114241681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62191591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAACATCACCTTCAACATTTC[A/G]ATAATTTTAACTTCTTCTGTTCCAG
Phenotype
MIM: 615060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCGB1D1 PubMed Links

Gene Details

Gene
SCGB1D1
Gene Name
secretoglobin family 1D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006552.1 Intron NP_006543.1

View Full Product Details