Product Details
- SNP ID
-
rs114749527
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:69641181 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGGGCTGTCGGCGCAGTAGCAGC[A/G]AGCAGCAGAGTCCGCACGCTCCGGC
- Phenotype
-
MIM: 168461
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCND1
PubMed Links
Gene Details
- Gene
- CCND1
- Gene Name
- cyclin D1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_053056.2 |
77 |
UTR 5 |
|
|
NP_444284.1 |
View Full Product Details