Product Details

SNP ID
rs114830141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59503765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACCCCCATGTACTTCCTGCTC[C/T]GCAATCTAGCCATCCTTGACATCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4D11 PubMed Links

Gene Details

Gene
OR4D11
Gene Name
olfactory receptor family 4 subfamily D member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004706.1 190 Missense Mutation CGC,TGC R64C NP_001004706.1

View Full Product Details