Product Details

SNP ID
rs114844139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:96353572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGATAAAATGTACAAACATGTCA[C/T]ATGATACAGAAAAACAAGAATCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC82 PubMed Links

Gene Details

Gene
CCDC82
Gene Name
coiled-coil domain containing 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318736.1 2058 UTR 3 NP_001305665.1
NM_001318737.1 2058 Intron NP_001305666.1
NM_024725.3 2058 UTR 3 NP_079001.2
XM_005274292.3 2058 UTR 3 XP_005274349.1
XM_006718911.3 2058 UTR 3 XP_006718974.1
XM_011542984.2 2058 UTR 3 XP_011541286.1
XM_011542987.2 2058 UTR 3 XP_011541289.1
XM_011542988.2 2058 UTR 3 XP_011541290.1
XM_017018305.1 2058 UTR 3 XP_016873794.1
XM_017018306.1 2058 UTR 3 XP_016873795.1
XM_017018307.1 2058 UTR 3 XP_016873796.1
XM_017018308.1 2058 UTR 3 XP_016873797.1
XM_017018309.1 2058 UTR 3 XP_016873798.1
XM_017018310.1 2058 UTR 3 XP_016873799.1
XM_017018311.1 2058 UTR 3 XP_016873800.1
XM_017018312.1 2058 UTR 3 XP_016873801.1

View Full Product Details