Product Details

SNP ID
rs114968130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94710766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCCATTTTTTGTCCCTCTTTTT[A/G]CTCCTAAATGCATTTCCAAAGGCTT
Phenotype
MIM: 614657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMOTL1 PubMed Links

Gene Details

Gene
AMOTL1
Gene Name
angiomotin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301007.1 Intron NP_001287936.1
NM_130847.2 Intron NP_570899.1
XM_005273798.4 Intron XP_005273855.1
XM_005273801.4 Intron XP_005273858.1
XM_006718772.3 Intron XP_006718835.1
XM_011542626.2 Intron XP_011540928.1
XM_011542628.2 Intron XP_011540930.1
Gene
LOC105369438
Gene Name
uncharacterized LOC105369438
There are no transcripts associated with this gene.

View Full Product Details