Product Details

SNP ID
rs115251370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61406672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTGTTTTCTAGGCAATATTGT[C/T]ACAGCATTTTCAAAGTTACCAAAAG
Phenotype
MIM: 613277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPSF7 PubMed Links

Gene Details

Gene
CPSF7
Gene Name
cleavage and polyadenylation specific factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136040.2 Intron NP_001129512.1
NM_001142565.1 Intron NP_001136037.1
NM_024811.3 Intron NP_079087.3
XM_005274298.4 Intron XP_005274355.1
XM_005274299.4 Intron XP_005274356.1
XM_005274303.4 Intron XP_005274360.1
XM_011545257.2 Intron XP_011543559.1
XM_011545258.2 Intron XP_011543560.1
XM_011545259.2 Intron XP_011543561.1
XM_011545260.2 Intron XP_011543562.1
XM_011545261.2 Intron XP_011543563.1
XM_011545262.2 Intron XP_011543564.1
XM_011545263.2 Intron XP_011543565.1
XM_017018345.1 Intron XP_016873834.1
Gene
TMEM216
Gene Name
transmembrane protein 216
There are no transcripts associated with this gene.

View Full Product Details