Product Details

SNP ID
rs115454950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73870288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAACACTTTTTATTATTAGTTAC[A/G]TATGTTTCATTCCCTCATTTATTTG
Phenotype
MIM: 608016 MIM: 611853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COA4 PubMed Links

Gene Details

Gene
COA4
Gene Name
cytochrome c oxidase assembly factor 4 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016565.2 3512 Intron NP_057649.2
XM_017017883.1 3512 UTR 3 XP_016873372.1
XM_017017884.1 3512 UTR 3 XP_016873373.1
Gene
MRPL48
Gene Name
mitochondrial ribosomal protein L48
There are no transcripts associated with this gene.

Gene
PAAF1
Gene Name
proteasomal ATPase associated factor 1
There are no transcripts associated with this gene.

View Full Product Details