Product Details

SNP ID
rs115523011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65617376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTGGGCCTGGACCTCAGCTTGT[C/T]CTCCTGTCCCTCTGCGAGCCAGTCC
Phenotype
MIM: 600050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K11 PubMed Links

Gene Details

Gene
MAP3K11
Gene Name
mitogen-activated protein kinase kinase kinase 11
There are no transcripts associated with this gene.

Gene
PCNX3
Gene Name
pecanex homolog 3 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032223.3 Intron NP_115599.2
XM_005273987.4 Intron XP_005274044.1
XM_005273990.4 Intron XP_005274047.1
XM_006718549.3 Intron XP_006718612.1
XM_006718551.3 Intron XP_006718614.1
XM_011545024.2 Intron XP_011543326.1
XM_011545025.2 Intron XP_011543327.1
XM_011545026.2 Intron XP_011543328.1
XM_011545027.1 Intron XP_011543329.1
XM_011545028.2 Intron XP_011543330.1

View Full Product Details