Product Details

SNP ID
rs116001129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68043647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGGACAGAAGATCCGCAAGATCA[C/T]GGACTGGTGAGTCACTGGGAACACC
Phenotype
MIM: 118491 MIM: 602141 MIM: 604592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
There are no transcripts associated with this gene.

Gene
MIR4691
Gene Name
microRNA 4691
There are no transcripts associated with this gene.

Gene
MIR6753
Gene Name
microRNA 6753
There are no transcripts associated with this gene.

Gene
NDUFS8
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
There are no transcripts associated with this gene.

Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006019.3 831 Missense Mutation ACG,ATG T236M NP_006010.2
NM_006053.3 831 Missense Mutation ACG,ATG T20M NP_006044.1
XM_005273709.3 831 Missense Mutation ACG,ATG T236M XP_005273766.1
XM_011544726.2 831 Missense Mutation ACG,ATG T236M XP_011543028.1
XM_017017089.1 831 Missense Mutation ACG,ATG T236M XP_016872578.1
XM_017017090.1 831 UTR 5 XP_016872579.1

View Full Product Details