Product Details

SNP ID
rs116711652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66412045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTACGAGCTGTGCCCTGATAGTA[A/G]CAAGGAGAGACTTGAAGCTGGAGCC
Phenotype
MIM: 608554
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPAS4 PubMed Links

Gene Details

Gene
NPAS4
Gene Name
neuronal PAS domain protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318804.1 Intron NP_001305733.1
NM_178864.3 Intron NP_849195.2
XM_017017537.1 Intron XP_016873026.1
XM_017017538.1 Intron XP_016873027.1
XM_017017539.1 Intron XP_016873028.1

View Full Product Details