Product Details

SNP ID
rs116864252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66363006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCCTCCTCTCAGGTGCCCACAA[C/T]TTCCCTGGCACCTTTTACCCTGGGT
Phenotype
MIM: 602110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724064 PubMed Links

Gene Details

Gene
LOC102724064
Gene Name
uncharacterized LOC102724064
There are no transcripts associated with this gene.

Gene
SLC29A2
Gene Name
solute carrier family 29 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300868.1 2181 UTR 3 NP_001287797.1
NM_001300869.1 2181 UTR 3 NP_001287798.1
NM_001532.2 2181 UTR 3 NP_001523.2
XM_011544974.1 2181 UTR 3 XP_011543276.1
XM_017017630.1 2181 UTR 3 XP_016873119.1
XM_017017631.1 2181 Intron XP_016873120.1
XM_017017632.1 2181 UTR 3 XP_016873121.1
XM_017017633.1 2181 Intron XP_016873122.1
XM_017017634.1 2181 Intron XP_016873123.1
XM_017017635.1 2181 UTR 3 XP_016873124.1
XM_017017636.1 2181 Intron XP_016873125.1
XM_017017637.1 2181 UTR 3 XP_016873126.1
XM_017017638.1 2181 UTR 3 XP_016873127.1

View Full Product Details